Assay Details
| Gene Symbol: Bud31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC058456.1 | - | 1 - 2 | 44 | 6887068 | 117 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
This assay is in the 5'UTR region of the BC058456.1 transcripts.
This assay is in the 5'UTR region of the BC058456.1 transcripts.
Set Membership: |
|

Bud31
Genomic Map