Assay Details
| Gene Symbol: Jakmip3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163277.1 | NP_001156749.1 | 3 - 4 | 956 | 56 | |
| XM_008760004.1 | XP_008758226.1 | 7 - 8 | 2051 | 56 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Jakmip3
Genomic Map