Assay Details
| Gene Symbol: Bles03 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024233.2 | NP_001019404.2 | 1 - 2 | 230 | 97 | |
| GenBank mRNA | CK596744.1 | - | 1 - 2 | 230 | 97 | |
| FQ233132.1 | - | 1 - 2 | 123 | 97 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Bles03
Genomic Map