Assay Details
| Gene Symbol: Pank1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106373.3 | NP_001099843.1 | 5 - 6 | 1105 | 98 | |
| XM_006231276.2 | XP_006231338.1 | 5 - 6 | 4408 | 98 | ||
| XM_006231277.2 | XP_006231339.1 | 4 - 5 | 4236 | 98 | ||
| XM_006231278.2 | XP_006231340.1 | 6 - 7 | 1756 | 98 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pank1
Genomic Map