Assay Details
| Gene Symbol: Ndufv1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006972.1 | NP_001006973.1 | 5 - 6 | 709 | 64 | |
| GenBank mRNA | BC083709.1 | - | 5 - 6 | 709 | 7190583 | 64 |
| FQ217076.1 | - | 5 - 6 | 766 | 64 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ndufv1
Genomic Map