Assay Details
| Gene Symbol: B3gat3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001128184.1 | NP_001121656.1 | 2 - 3 | 432 | 68 | |
| GenBank mRNA | BC166510.1 | - | 2 - 3 | 310 | 7127430 | 68 |
| BC168710.1 | - | 2 - 3 | 413 | 8364370 | 68 | |
More Information
Updated Mapping Information
Set Membership: |
|

B3gat3
Genomic Map