Assay Details
| Gene Symbol: Vegfb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053549.1 | NP_446001.1 | 2 - 3 | 354 | 90 | |
| XM_006230910.2 | XP_006230972.1 | 2 - 3 | 451 | 90 | ||
| XM_006230911.2 | XP_006230973.1 | 2 - 3 | 452 | 90 | ||
| XM_006230912.2 | XP_006230974.1 | 2 - 3 | 442 | 90 | ||
| GenBank mRNA | BC076395.1 | - | 2 - 3 | 354 | 7109119 | 90 |
More Information
Updated Mapping Information
Set Membership: |
|

Vegfb
Genomic Map