Assay Details
| Gene Symbol: Vegfb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006230910.2 | XP_006230972.1 | 6 - 7 | 995 | 91 | |
| XM_006230912.2 | XP_006230974.1 | 6 - 7 | 885 | 91 | ||
More Information
Set Membership: |
|

Vegfb
Genomic Map