Assay Details
| Gene Symbol: Trpt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106331.1 | NP_001099801.1 | 5 - 6 | 879 | 71 | |
| XM_006230761.2 | XP_006230823.1 | 5 - 6 | 569 | 71 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Trpt1
Genomic Map