Assay Details
| Gene Symbol: Lipk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106374.1 | NP_001099844.1 | 3 - 4 | 328 | 64 | |
| XM_008760325.1 | XP_008758547.1 | 4 - 5 | 336 | 64 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Lipk
Genomic Map