Assay Details
| Gene Symbol: Rras | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001108481.1 | NP_001101951.1 | 1 - 2 | 168 | 84 | |
| GenBank mRNA | FQ211166.1 | - | 1 - 2 | 184 | 84 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rras
Genomic Map