Assay Details
| Gene Symbol: Etfb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004220.1 | NP_001004220.1 | 2 - 3 | 248 | 75 | |
| GenBank mRNA | BC079351.1 | - | 2 - 3 | 248 | 7108112 | 75 |
More Information
Updated Mapping Information
Set Membership: |
|

Etfb
Genomic Map