Assay Details
| Gene Symbol: Akt1s1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001106259.3 | NP_001099729.2 | 1 - 2 | 115 | 74 | |
| GenBank mRNA | FM032217.1 | - | 1 - 2 | 115 | 74 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the NM_001106259.3 transcripts.
Set Membership: |
|

Akt1s1
Genomic Map