Assay Details
| Gene Symbol: Hsd17b14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001191111.1 | NP_001178040.1 | 7 - 8 | 541 | 69 | |
More Information
Updated Mapping Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hsd17b14| Gene Symbol: Hsd17b14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001191111.1 | NP_001178040.1 | 7 - 8 | 541 | 69 | |
Set Membership: |
|