Assay Details
| Gene Symbol: Mark4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001191071.1 | NP_001178000.1 | 1 - 2 | 78 | 67 | |
| XM_006228472.2 | XP_006228534.1 | 1 - 2 | 118 | 67 | ||
| XM_008758981.1 | XP_008757203.1 | 1 - 2 | 118 | 67 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mark4
Genomic Map