Assay Details
| Gene Symbol: Ccdc155 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006229199.2 | XP_006229261.1 | 10 - 11 | 1479 | 62 | |
| XM_006229201.2 | XP_006229263.1 | 10 - 11 | 1168 | 62 | ||
More Information
Set Membership: |
|

Ccdc155
Genomic Map