Assay Details
| Gene Symbol: Hipk4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024776.1 | NP_001019947.1 | 1 - 2 | 722 | 65 | |
| GenBank mRNA | BC098070.1 | - | 1 - 2 | 722 | 7455473 | 65 |
More Information
Updated Mapping Information
Set Membership: |
|

Hipk4
Genomic Map