Assay Details
| Gene Symbol: Hipk4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024776.1 | NP_001019947.1 | 2 - 3 | 1076 | 89 | |
| GenBank mRNA | BC098070.1 | - | 2 - 3 | 1076 | 7455473 | 89 |
More Information
Updated Mapping Information
Set Membership: |
|

Hipk4
Genomic Map