Assay Details
| Gene Symbol: Orm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053288.2 | NP_445740.1 | 1 - 2 | 152 | 112 | |
| GenBank mRNA | FQ209739.1 | - | 1 - 2 | 154 | 112 | |
| FQ210338.1 | - | 1 - 2 | 154 | 112 | ||
| FQ218200.1 | - | 1 - 2 | 157 | 112 | ||
| FQ218302.1 | - | 1 - 2 | 154 | 112 | ||
| FQ219945.1 | - | 1 - 2 | 158 | 112 | ||
| FQ220896.1 | - | 1 - 2 | 158 | 112 | ||
| FQ221095.1 | - | 1 - 2 | 153 | 112 | ||
| FQ222215.1 | - | 1 - 2 | 154 | 112 | ||
| FQ222829.1 | - | 1 - 2 | 154 | 112 | ||
| FQ223035.1 | - | 1 - 2 | 154 | 112 | ||
| FQ228371.1 | - | 1 - 2 | 158 | 112 | ||
| FQ228464.1 | - | 1 - 2 | 152 | 112 | ||
| FQ228466.1 | - | 1 - 2 | 154 | 112 | ||
| FQ228489.1 | - | 1 - 2 | 154 | 112 | ||
| FQ228584.1 | - | 1 - 2 | 154 | 112 | ||
| FQ228800.1 | - | 1 - 2 | 154 | 112 | ||
| FQ229026.1 | - | 1 - 2 | 154 | 112 | ||
| FQ229654.1 | - | 1 - 2 | 152 | 112 | ||
| FQ230009.1 | - | 1 - 2 | 158 | 112 | ||
| J00696.1 | - | 1 - 2 | 153 | 112 | ||
| V01216.1 | - | 1 - 2 | 152 | 112 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Orm1
Genomic Map