Assay Details
| Gene Symbol: Cdc26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001013240.2 | NP_001013258.1 | 2 - 3 | 171 | 177 | |
| NM_001267054.1 | NP_001253983.1 | 2 - 3 | 99 | 105 | ||
| GenBank mRNA | AY156922.1 | - | 2 - 3 | 167 | 177 | |
| BC086343.1 | - | 2 - 3 | 160 | 7301739 | 177 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AY156922.1, BC086343.1, NM_001013240.2, NM_001267054.1 transcripts.
Set Membership: |
|

Cdc26
Genomic Map