Assay Details
| Gene Symbol: Bglap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013414.1 | NP_038200.1 | 2 - 3 | 153 | 81 | |
| XM_006232594.2 | XP_006232656.1 | 3 - 4 | 3210 | 81 | ||
| GenBank mRNA | M11777.1 | - | 2 - 3 | 133 | 81 | |
| X04141.1 | - | 2 - 3 | 152 | 81 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Bglap
Genomic Map