Assay Details
| Gene Symbol: Gba | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001127639.1 | NP_001121111.1 | 4 - 5 | 535 | 71 | |
| GenBank mRNA | BC166732.1 | - | 4 - 5 | 535 | 7934870 | 71 |
More Information
Updated Mapping Information
Set Membership: |
|

Gba
Genomic Map