Assay Details
| Gene Symbol: Tmem79 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033896.1 | NP_001029068.1 | 2 - 3 | 963 | 105 | |
| XM_006232637.2 | XP_006232699.1 | 2 - 3 | 1017 | 105 | ||
| XM_008761149.1 | XP_008759371.1 | 1 - 2 | 1482 | 105 | ||
| GenBank mRNA | BC101913.1 | - | 2 - 3 | 963 | 7936201 | 105 |
More Information
Updated Mapping Information
Set Membership: |
|

Tmem79
Genomic Map