Assay Details
| Gene Symbol: Hdgf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053707.2 | NP_446159.1 | 2 - 3 | 440 | 110 | |
| GenBank mRNA | AF389348.1 | - | 2 - 3 | 169 | 110 | |
| AF448810.1 | - | 2 - 3 | 169 | 110 | ||
| BC070943.1 | - | 2 - 3 | 440 | 7103046 | 110 | |
| FQ231256.1 | - | 3 - 4 | 203 | 110 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Hdgf
Genomic Map