Assay Details
| Gene Symbol: Nes | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012987.1 | NP_037119.1 | 3 - 4 | 1113 | 79 | |
| XM_006232600.2 | XP_006232662.1 | 3 - 4 | 1431 | 79 | ||
| GenBank mRNA | AF538924.1 | - | 3 - 4 | 986 | 79 | |
| M34384.1 | - | 3 - 4 | 1113 | 79 | ||
| Gene Symbol: LOC100910255 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006232557.2 | XP_006232619.1 | 3 - 4 | 1432 | 79 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Nes
Genomic Map