Assay Details
| Gene Symbol: Tbp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004198.1 | NP_001004198.1 | 4 - 5 | 791 | 75 | |
| XM_006227980.2 | XP_006228042.1 | 4 - 5 | 734 | 75 | ||
| XM_008758747.1 | XP_008756969.1 | 4 - 5 | 734 | 75 | ||
| XM_008758748.1 | XP_008756970.1 | 4 - 5 | 942 | 75 | ||
| GenBank mRNA | BC081939.1 | - | 4 - 5 | 791 | 7115389 | 75 |
More Information
Updated Mapping Information
Set Membership: |
|

Tbp
Genomic Map