Assay Details
| Gene Symbol: Fam71f1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024324.1 | NP_001019495.1 | 1 - 2 | 287 | 89 | |
| XM_006236223.2 | XP_006236285.1 | 3 - 4 | 649 | 89 | ||
| XM_006236224.2 | XP_006236286.1 | 3 - 4 | 649 | 89 | ||
| XM_006236225.2 | XP_006236287.1 | 3 - 4 | 649 | 89 | ||
| XM_008762818.1 | XP_008761040.1 | 3 - 4 | 649 | 89 | ||
| GenBank mRNA | BC079297.1 | - | 1 - 2 | 287 | 7134497 | 89 |
More Information
Set Membership: |
|

Fam71f1
Genomic Map