Assay Details
| Gene Symbol: Ubn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001134553.1 | NP_001128025.1 | 12 - 13 | 2221 | 77 | |
| XM_006236306.2 | XP_006236368.1 | 12 - 13 | 3310 | 77 | ||
| XM_006236307.2 | XP_006236369.1 | 12 - 13 | 3310 | 77 | ||
| XM_006236308.2 | XP_006236370.1 | 12 - 13 | 3304 | 77 | ||
| XM_006236311.2 | XP_006236373.1 | 12 - 13 | 3310 | 77 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ubn2
Genomic Map