Assay Details
| Gene Symbol: Gstk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_181371.2 | NP_852036.1 | 2 - 3 | 187 | 91 | |
| GenBank mRNA | FJ179400.1 | - | 2 - 3 | 153 | 91 | |
| FQ211030.1 | - | 2 - 3 | 205 | 91 | ||
| FQ214179.1 | - | 2 - 3 | 270 | 91 | ||
| FQ220456.1 | - | 2 - 3 | 243 | 91 | ||
| S83436.1 | - | 2 - 3 | 187 | 91 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Gstk1
Genomic Map