Assay Details
| Gene Symbol: Tcaf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003749739.3 | XP_003749787.1 | 7 - 8 | 2672 | 65 | |
| XM_008762928.1 | XP_008761150.1 | 7 - 8 | 3001 | 65 | ||
| XM_342675.6 | XP_342676.4 | 7 - 8 | 3001 | 65 | ||
| GenBank mRNA | FQ211472.1 | - | 7 - 8 | 2639 | 65 | |
More Information
Set Membership: |
|

Tcaf1
Genomic Map