Assay Details
| Gene Symbol: Creb3l2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012188.1 | NP_001012188.1 | 6 - 7 | 1249 | 115 | |
| GenBank mRNA | AY546001.1 | - | 6 - 7 | 1249 | 115 | |
More Information
Updated Mapping Information
Set Membership: |
|

Creb3l2
Genomic Map