Assay Details
| Gene Symbol: Creb3l2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012188.1 | NP_001012188.1 | 8 - 9 | 1374 | 63 | |
| GenBank mRNA | AY546001.1 | - | 8 - 9 | 1374 | 63 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
chondrocyte differentiation
transcription, DNA-templated
transcription from RNA polymerase II promoter
ER to Golgi vesicle-mediated transport
endoplasmic reticulum unfolded protein response
response to endoplasmic reticulum stress
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
cartilage development
RNA polymerase II core promoter proximal region sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
cAMP response element binding
transcription regulatory region DNA binding
chondrocyte differentiation
transcription, DNA-templated
transcription from RNA polymerase II promoter
ER to Golgi vesicle-mediated transport
endoplasmic reticulum unfolded protein response
response to endoplasmic reticulum stress
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
cartilage development
RNA polymerase II core promoter proximal region sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
cAMP response element binding
transcription regulatory region DNA binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P083788 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 464369 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 929173 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 625546 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 714429 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 262142 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 544353 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 7805 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 702223 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 104673 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 504440 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 729233 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 262271 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 654381 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 291965 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 778045 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 270558 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 245760 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 928189 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 536445 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 144119 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 768237 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 867311 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 127067 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 115578 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 187237 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 647489 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 311542 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 85331 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 47787 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 518324 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 147464 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 492541 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 843332 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 857605 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 873088 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 101698 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 42830 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 308395 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 148913 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 295109 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 849981 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 121669 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 529920 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 590005 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 790299 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 367074 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 66669 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 634019 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 433294 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 580782 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 75709 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 364028 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 228482 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 80222 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 552653 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 806626 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 690041 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 562438 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 79752 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 687517 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 245981 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 46362 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 857340 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 402878 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 916292 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 238661 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 588454 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 716755 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 199190 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 553108 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 685143 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 636819 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 119619 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 429589 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 587152 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 818159 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 589416 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 397315 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 102154 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 451429 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 432198 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 697101 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 43799 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 643017 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 887797 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 592074 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 310699 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 157230 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 58726 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 63170 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 145893 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 9256 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 75612 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 13209 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 13268 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 96547 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 62431 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 167292 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 152295 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 128377 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 34530 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 134533 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 80221 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P083788 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P083788 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P083788 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P144980 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 5584 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 29828 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 51848 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 51781 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 43470 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 31304 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 101756 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 64168 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10861897 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10861891 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10861893 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10861899 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10861901 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10861896 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10861902 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10861892 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10861903 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10861894 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10861904 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10861895 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10861890 | - | - |
| Rat Genome 230 2.0 Array | 1379565_at | - | - |
| Rat Genome 230 2.0 Array | 1379565_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1379565_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1379565_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0400019157.rn.1 | TC0400002700.rn.1 | RTA TC |
| Rat Expression Set 230 | 1379565_at | - | - |
| Rat Expression Set 230 | 1379565_PM_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10861890 | - | - |
| Rat Genome U34 Set | rc_AA946426_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21197500 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1376830 | - | - |

Creb3l2
Genomic Map