Assay Details
| Gene Symbol: Rbm28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107850.1 | NP_001101320.1 | 1 - 2 | 200 | 90 | |
| XM_006236190.2 | XP_006236252.1 | 2 - 3 | 376 | 90 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rbm28
Genomic Map