Assay Details
| Gene Symbol: Lrrc45 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001127571.1 | NP_001121043.1 | 6 - 7 | 798 | 59 | |
| XM_006247924.1 | XP_006247986.1 | 6 - 7 | 853 | 59 | ||
| GenBank mRNA | BC161940.1 | - | 684 | 9023431 | 59 | |
More Information
Updated Mapping Information
Set Membership: |
|

Lrrc45
Genomic Map