Assay Details
| Gene Symbol: Irf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012591.1 | NP_036723.1 | 3 - 4 | 66 | ||
| GenBank mRNA | BC076382.1 | - | 3 - 4 | 302 | 7099391 | 66 |
| M34253.1 | - | 3 - 4 | 66 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Irf1
Genomic Map