Assay Details
| Gene Symbol: Il4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201270.1 | NP_958427.1 | 2 - 3 | 214 | 63 | |
| GenBank mRNA | X16058.2 | - | 2 - 3 | 214 | 63 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: Il4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201270.1 | NP_958427.1 | 2 - 3 | 214 | 63 | |
| GenBank mRNA | X16058.2 | - | 2 - 3 | 214 | 63 | |
Set Membership: |
|