Assay Details
| Gene Symbol: Plk4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001107669.1 | NP_001101139.1 | 8 - 9 | 1980 | 83 | |
| XM_006232299.2 | XP_006232361.1 | 9 - 10 | 2158 | 83 | ||
| XM_006232300.1 | XP_006232362.1 | 8 - 9 | 1863 | 83 | ||
| GenBank mRNA | BC166450.1 | - | 9 - 10 | 2098 | 8371068 | 83 |
| FQ234784.1 | - | 5 - 6 | 1030 | 83 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Plk4
Genomic Map