Assay Details
| Gene Symbol: Usp42 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105909.1 | NP_001099379.1 | 4 - 5 | 651 | 63 | |
| XM_008768948.1 | XP_008767170.1 | 4 - 5 | 661 | 63 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Usp42
Genomic Map