Assay Details
| Gene Symbol: Mmd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037217.1 | NP_001032294.1 | 2 - 3 | 299 | 62 | |
| GenBank mRNA | BC161816.1 | - | 2 - 3 | 277 | 7320318 | 62 |
| CO400908.1 | - | 2 - 3 | 276 | 62 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mmd2
Genomic Map