Assay Details
| Gene Symbol: Mmd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037217.1 | NP_001032294.1 | 4 - 5 | 542 | 74 | |
| GenBank mRNA | BC161816.1 | - | 4 - 5 | 520 | 7320318 | 74 |
| CO400908.1 | - | 4 - 5 | 519 | 74 | ||
| CO404759.1 | - | 1 - 2 | 82 | 74 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mmd2
Genomic Map