Assay Details
| Gene Symbol: Gpc4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014108.1 | NP_001014130.1 | 3 - 4 | 1227 | 111 | |
| XM_006257580.2 | XP_006257642.1 | 4 - 5 | 825 | 111 | ||
| GenBank mRNA | BC081962.1 | - | 3 - 4 | 1227 | 7124728 | 111 |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Rat Genome Microarray 4x44K v2 | A_42_P547976 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 264147 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 337074 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 76443 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 972589 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 227413 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 678033 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 351954 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 688726 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 639985 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 278118 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 461719 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 845535 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 302056 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 150290 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 491727 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 132828 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 640507 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 881414 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 754270 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 135761 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 424490 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 130261 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 508415 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 153335 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 24607 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 384669 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 342338 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 622893 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 6947 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 82914 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 947255 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 124359 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 337675 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 852663 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 536556 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 253067 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 565404 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 275291 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 702341 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 398266 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 400339 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 566257 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 913287 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 881832 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 946151 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 816927 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 945267 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 686039 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 8994 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 169661 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 789278 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 186361 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 107948 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 865578 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 155815 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 795883 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 22973 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 552115 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 542096 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 95888 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 62717 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 757796 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 74508 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 119393 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 452267 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 894209 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 708478 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 769874 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 84554 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 968177 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 424370 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 138777 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 495538 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 600323 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 261387 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 869455 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 967646 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 187395 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 325533 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 899671 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 668809 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 672730 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 756937 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 609539 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 256748 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 648984 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 708405 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 1x1M | 463842 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 169418 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 131554 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 87399 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 174109 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 51984 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 144744 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 152106 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 52836 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 3480 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 105749 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 178900 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 155009 | - | - |
| Agilent SurePrint G3 Rat CGH Microarray 4x180K | 50830 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_42_P547976 | - | - |
| Agilent Whole Rat Genome Microarray 4x44K v3 | A_42_P547976 | - | - |
| Agilent SurePrint G3 Rat GE 8x60K Microarray | A_42_P547976 | - | - |
| SurePrint G3 Rat Gene Expression Microarray G4131A | A_43_P21658 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 65553 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 99229 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 54652 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 8642 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 43161 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 52234 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 66208 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 39437 | - | - |
| Agilent Rat Genome CGH Microarray 105A (G4436A) | 58747 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Rat Gene 1.0 ST Array | 10939746 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10939745 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10939747 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10939749 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10939752 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10939748 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10939751 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10939750 | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10939753 | - | - |
| Affymetrix Rat Gene 1.1 ST Array | 10939744 | - | - |
| Rat Genome 230 2.0 Array | 1392888_at | - | - |
| Rat Genome 230 2.0 Array | 1392888_PM_at | - | - |
| Affymetrix HT Rat Focus Array | 1392888_at | - | - |
| Affymetrix HT Rat Focus Array | 1392888_PM_at | - | - |
| Affymetrix Rat Clariom S Assay | PSR0X00015653.rn.1 | TC0X00002486.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0X00015651.rn.1 | TC0X00002486.rn.1 | RTA TC |
| Affymetrix Rat Clariom S Assay | PSR0X00015652.rn.1 | TC0X00002486.rn.1 | RTA TC |
| Rat Expression Set 230 | 1392888_PM_at | - | - |
| Rat Expression Set 230 | 1392888_at | - | - |
| Affymetrix Rat Gene 1.0 ST Array | 10939744 | - | - |
| Rat Genome U34 Set | rc_AI071251_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Rat Genome Survey Microarray | 21878463 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina ratRef-12 v1.0 expression beadchip | ILMN_1650537 | - | - |

Gpc4
Genomic Map