Assay Details
| Gene Symbol: Rab4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017355.1 | NP_059051.1 | 4 - 5 | 357 | 57 | |
| XM_008759181.1 | XP_008757403.1 | 3 - 4 | 576 | 57 | ||
| XM_008759182.1 | XP_008757404.1 | 3 - 4 | 576 | 57 | ||
| GenBank mRNA | FQ227674.1 | - | 4 - 5 | 389 | 57 | |
| X78605.1 | - | 4 - 5 | 357 | 57 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rab4b
Genomic Map