Assay Details
| Gene Symbol: Kcnc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053997.4 | NP_446449.2 | 2 - 3 | 2201 | 67 | |
| XM_006228973.2 | XP_006229035.1 | 2 - 3 | 2395 | 67 | ||
| XM_008759308.1 | XP_008757530.1 | 2 - 3 | 2395 | 67 | ||
| GenBank mRNA | AY179603.1 | - | 2 - 3 | 2201 | 67 | |
| AY179604.1 | - | 2 - 3 | 2201 | 67 | ||
| AY822673.1 | - | 2 - 3 | 2201 | 67 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Kcnc3
Genomic Map