Assay Details
| Gene Symbol: Kcnc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006228965.2 | XP_006229027.1 | 2 - 3 | 2395 | 62 | |
| XM_006228971.2 | XP_006229033.1 | 2 - 3 | 2395 | 62 | ||
| XM_008759306.1 | XP_008757528.1 | 2 - 3 | 2395 | 62 | ||
| XM_008759309.1 | XP_008757531.1 | 2 - 3 | 2395 | 62 | ||
| GenBank mRNA | M84210.1 | - | 2 - 3 | 2201 | 62 | |
More Information
Set Membership: |
|

Kcnc3
Genomic Map