Assay Details
| Gene Symbol: Zbtb32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109517.1 | NP_001102987.1 | 2 - 3 | 979 | 76 | |
| XM_006228793.2 | XP_006228855.1 | 3 - 4 | 1651 | 76 | ||
| XM_008759188.1 | XP_008757410.1 | 3 - 4 | 2004 | 76 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Zbtb32
Genomic Map