Assay Details
Gene Symbol: Cyp2b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001134844.1 | NP_001128316.1 | 985 | 101 | ||
XM_008765491.1 | XP_008763713.1 | 1028 | 101 | |||
XM_008765492.1 | XP_008763714.1 | 1004 | 101 | |||
XM_008765493.1 | XP_008763715.1 | 1390 | 101 | |||
GenBank mRNA | BC166502.1 | - | 985 | 7122207 | 101 | |
J00719.1 | - | 953 | 101 | |||
M37134.1 | - | 953 | 101 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
