Assay Details
| Gene Symbol: Spint2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082549.1 | NP_001076018.1 | 2 - 3 | 558 | 62 | |
| GenBank mRNA | DN934634.1 | - | 2 - 3 | 587 | 62 | |
More Information
Set Membership: |
|

Spint2
Genomic Map