Assay Details
Gene Symbol: Ftl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022500.4 | NP_071945.3 | 1 - 2 | 307 | 60 | |
GenBank mRNA | BC061525.1 | - | 1 - 2 | 311 | 6921603 | 60 |
BC086583.1 | - | 1 - 2 | 288 | 7307718 | 60 | |
BC088756.1 | - | 1 - 2 | 288 | 7307747 | 60 | |
FQ209407.1 | - | 1 - 2 | 307 | 60 | ||
FQ209694.1 | - | 1 - 2 | 307 | 60 | ||
FQ209816.1 | - | 1 - 2 | 307 | 60 | ||
FQ218577.1 | - | 1 - 2 | 307 | 60 | ||
FQ219483.1 | - | 1 - 2 | 307 | 60 | ||
FQ219685.1 | - | 1 - 2 | 307 | 60 | ||
FQ219809.1 | - | 1 - 2 | 307 | 60 | ||
FQ219930.1 | - | 1 - 2 | 306 | 60 | ||
FQ220095.1 | - | 1 - 2 | 307 | 60 | ||
FQ220123.1 | - | 1 - 2 | 307 | 60 | ||
FQ220243.1 | - | 1 - 2 | 304 | 60 | ||
FQ220651.1 | - | 1 - 2 | 307 | 60 | ||
FQ220814.1 | - | 1 - 2 | 307 | 60 | ||
FQ220884.1 | - | 1 - 2 | 307 | 60 | ||
FQ221898.1 | - | 1 - 2 | 307 | 60 | ||
FQ223227.1 | - | 1 - 2 | 307 | 60 | ||
FQ226019.1 | - | 1 - 2 | 307 | 60 | ||
FQ226254.1 | - | 1 - 2 | 263 | 60 | ||
FQ226458.1 | - | 1 - 2 | 306 | 60 | ||
FQ226986.1 | - | 1 - 2 | 303 | 60 | ||
FQ227401.1 | - | 1 - 2 | 307 | 60 | ||
FQ227800.1 | - | 1 - 2 | 307 | 60 | ||
FQ227843.1 | - | 1 - 2 | 307 | 60 | ||
FQ228050.1 | - | 1 - 2 | 307 | 60 | ||
FQ228585.1 | - | 1 - 2 | 307 | 60 | ||
FQ229176.1 | - | 1 - 2 | 307 | 60 | ||
FQ229284.1 | - | 1 - 2 | 307 | 60 | ||
FQ229370.1 | - | 1 - 2 | 307 | 60 | ||
FQ229673.1 | - | 1 - 2 | 305 | 60 | ||
FQ229757.1 | - | 1 - 2 | 307 | 60 | ||
FQ230445.1 | - | 1 - 2 | 307 | 60 | ||
FQ230659.1 | - | 1 - 2 | 303 | 60 | ||
FQ231322.1 | - | 1 - 2 | 307 | 60 | ||
FQ231373.1 | - | 1 - 2 | 307 | 60 | ||
FQ231464.1 | - | 1 - 2 | 307 | 60 | ||
FQ231570.1 | - | 1 - 2 | 307 | 60 | ||
FQ231615.1 | - | 1 - 2 | 307 | 60 | ||
FQ231781.1 | - | 1 - 2 | 303 | 60 | ||
FQ231957.1 | - | 1 - 2 | 307 | 60 | ||
FQ232031.1 | - | 1 - 2 | 307 | 60 | ||
FQ232167.1 | - | 1 - 2 | 307 | 60 | ||
FQ232317.1 | - | 1 - 2 | 263 | 60 | ||
FQ232517.1 | - | 1 - 2 | 307 | 60 | ||
FQ233772.1 | - | 1 - 2 | 307 | 60 | ||
FQ233996.1 | - | 1 - 2 | 307 | 60 | ||
FQ234074.1 | - | 1 - 2 | 307 | 60 | ||
FQ234603.1 | - | 1 - 2 | 307 | 60 | ||
FQ234647.1 | - | 1 - 2 | 307 | 60 | ||
FQ235044.1 | - | 1 - 2 | 307 | 60 | ||
FQ235250.1 | - | 1 - 2 | 307 | 60 | ||
K01930.1 | - | 1 - 2 | 232 | 60 | ||
L01122.1 | - | 1 - 2 | 260 | 60 |
Gene Symbol: LOC100360087 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002729553.4 | XP_002729599.1 | 1 | 322 | 60 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
