Assay Details
| Gene Symbol: LOC100910336 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003750647.3 | XP_003750695.1 | 2 - 3 | 180 | 64 | |
| XM_008766901.1 | XP_008765123.1 | 2 - 3 | 92 | 64 | ||
| Gene Symbol: LOC103690015 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002728672.4 | XP_002728718.4 | 1 | 96 | 64 | |
| Gene Symbol: Rps19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037346.2 | NP_001032423.1 | 2 - 3 | 125 | 64 | |
| XM_006228411.1 | XP_006228473.1 | 2 - 3 | 92 | 64 | ||
| GenBank mRNA | BC087641.1 | - | 2 - 3 | 97 | 7308408 | 64 |
| CB720565.1 | - | 2 - 3 | 125 | 64 | ||
| FQ221309.1 | - | 2 - 3 | 107 | 64 | ||
| FQ221510.1 | - | 2 - 3 | 108 | 64 | ||
| FQ224282.1 | - | 2 - 3 | 108 | 64 | ||
| FQ229780.1 | - | 2 - 3 | 108 | 64 | ||
| X51707.1 | - | 2 - 3 | 107 | 64 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

LOC100910336
Genomic Map