Assay Details
| Gene Symbol: Kctd15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109141.1 | NP_001102611.1 | 1 - 2 | 264 | 75 | |
| XM_006228859.2 | XP_006228921.1 | 2 - 3 | 115 | 75 | ||
| XM_006228860.2 | XP_006228922.1 | 2 - 3 | 368 | 75 | ||
| XM_008759178.1 | XP_008757400.1 | 2 - 3 | 190 | 75 | ||
| GenBank mRNA | BC168920.1 | - | 1 - 2 | 208 | 9100405 | 75 |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC168920.1, NM_001109141.1, XM_006228859.2, XM_006228860.2, XM_008759178.1 transcripts.
Set Membership: |
|

Kctd15
Genomic Map